Skip to Content
Merck
All Photos(2)

Key Documents

792004

Sigma-Aldrich

Copper(II) oxide

nanorod, diam. × L 10-12 nm × 75-100 nm

Synonym(s):

Copper(II) oxide, Cupric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

nanorod

diam. × L

10-12 nm × 75-100 nm

surface area

600-100 m2/g

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Nanoscale Copper(II) Oxide has been studied as photocatalysts, sensors, lubricant additives and batteries.
  • Nanorods of cupric oxide have also shown advantages as oxidizing agents in high speed chemical reactions over traditional cupric oxide nanoparticles.
  • Copper(II) oxide is a promising p-type oxide material although with a small band gap.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasound assisted shape regulation of CuO nanorods in ionic liquids and their use as energy efficient lubricant additives.
Gusain, et al.
Journal of Material Chemistry A, 1(18), 5612-5619 (2013)
Facile synthesis of CuO nanorod for lithium storage application.
Yang, et al.
Materials Letters, 90, 4-7 (2013)
A novel combustion method to prepare CuO nanorods and its antimicrobial and photocatalytic activities.
Christy, et al.
Powder Technology, 235, 783-786 (2013)
Combustion characteristics of novel hybrid nanoenergetic formulations.
Thiruvengadathan et al.
Combustion and Flame, 158(5), 964-978 (2011)
Synthesis and ethanol sensing properties of CuO nanorods coated with In2O3.
Park, et al.
Ceramics International, 39(5), 5255-5262 (2013)

Articles

Electronically, it behaves as a wide band gap (3.2 eV) semiconductor and exhibits memristor properties.2 Optically, TiO2 has high opacity with a very high refractive index3 (>2.4), and it exhibits strong absorbance in the UV range.

The past several decades have seen major advancements in the synthesis of metal nanomaterials. Most recently, controlled synthesis has become versatile enough to regulate the exact number of atoms and ligands of very small metal nanoparticles, referred to as “clusters”.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service