Skip to Content
Merck
All Photos(1)

Key Documents

772569

Sigma-Aldrich

Polystyrene, DDMAT terminated

average Mn 10,000, PDI ≤1.1

Synonym(s):

Poly(styrene) RAFT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7O2(C8H8)nC13H25S3
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

mol wt

average Mn 10,000

transition temp

Tm 378-383 °C

PDI

≤1.1

Looking for similar products? Visit Product Comparison Guide

Application

Poly(styrene) is a hydrophobic polymer used in many applications. The end group functionality contains a carboxylic acid (COOH) as well as the 2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid RAFT agent moiety. This will allow for additional polymerization to generate a diblock copolymer. This RAFT agent is especially suited for the polymerization of styrene; acrylate and acrylamide monomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service