Skip to Content
Merck
All Photos(1)

Key Documents

747971

Sigma-Aldrich

Gold nanorods

10 nm diameter, silica coated, λmax, 780 nm, dispersion in H2O

Synonym(s):

Au nanorods, Gold nanorod

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

material

silica

form

dispersion in H2O
nanorod

composition

, 7.8-8.2 x 10^11 particles/mL

concentration

>30 μg/mL in H2O

diameter

10 nm

λmax

780 nm

storage temp.

2-8°C

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Our silica coated gold nanorods are highly stable and suitable for applications including; photothermal therapy; fluorescence enhancement; plasmonic arrays; and photoacoustic amplifiers.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun-Sheng Chen et al.
Nano letters, 11(2), 348-354 (2011-01-20)
Photoacoustic signal generation by metal nanoparticles relies on the efficient conversion of light to heat, its transfer to the environment, and the production of pressure transients. In this study we demonstrate that a dielectric shell has a strong influence on
Liu; Si-Yun;
The Journal of Physical Chemistry C, 117(20), 10636-10642 (2013)
Stéphanie Vial et al.
Langmuir : the ACS journal of surfaces and colloids, 23(8), 4606-4611 (2007-03-21)
A systematic study of the optical effects derived from plasmon coupling in mono- and multilayers of gold nanorods is presented. The monolayers were prepared using the standard polyelectrolyte-assisted layer-by-layer (LbL) method and gold nanorods coated with either poly(N-vinyl pyrrolidone) or
Ina Weidenfeld et al.
Nature communications, 10(1), 5056-5056 (2019-11-09)
Macrophages are one of the most functionally-diverse cell types with roles in innate immunity, homeostasis and disease making them attractive targets for diagnostics and therapy. Photo- or optoacoustics could provide non-invasive, deep tissue imaging with high resolution and allow to
Sudipta Mallick et al.
Journal of nanoscience and nanotechnology, 13(5), 3223-3229 (2013-07-19)
Due to their efficient conversion of absorbed light energy to heat gold nanorods have been proved to be an amazing tool for minimally invasive photo-thermal cancer therapy. The present in vitro study demonstrates the ability of silica coated Au nanorods

Articles

In the past few decades, research and commercial use of gold nanoparticles has rapidly expanded into fields extending from biomedical engineering to photovoltaics. Most applications take advantage of the excellent optical properties of gold nanoparticles, which can be fine-tuned by varying their shape and size.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service