Skip to Content
Merck
All Photos(1)

Key Documents

743828

Sigma-Aldrich

1,3-Butadiene

≥99.6%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH=CH2
CAS Number:
Molecular Weight:
54.09
Beilstein:
605258
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.9 (15 °C, vs air)

vapor pressure

1863 mmHg ( 21 °C)

Assay

≥99.6%

form

gas

autoignition temp.

788 °F

expl. lim.

12 %

bp

−4.5 °C (lit.)

mp

−109 °C (lit.)

density

0.62 g/mL at 20 °C (lit.)

SMILES string

C=CC=C

InChI

1S/C4H6/c1-3-4-2/h3-4H,1-2H2

InChI key

KAKZBPTYRLMSJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Butadiene can be used as a starting material:
  • In the synthesis of 1-butene by selective hydrogenation reaction using various supported catalysts.
  • Along with acrylic acid in the multistep synthesis of oseltamivir.
  • For 1,4-cis-polymerization by Gd complex/iBu3Al catalyst system.
  • In combination with alcohols for the synthesis of alkyl octadienyl ethers by telomerization reaction using palladium−carbene catalysts.

Biochem/physiol Actions

Environmental carcinogen. Induces cardiac hemangiosarcomas in mice.

Other Notes

For 743828-250G-EU packaging: 500 mL of aluminium cylinder equipped with brass valve.

Recommended products

For 743828-250G-EU: outlet fitting: DIN 477-1 no. 1, Z742160 or Z742161 are the recommended regulators.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Gas 1A - Muta. 1B - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-104.8 °F - closed cup

Flash Point(C)

-76 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melike Babucci et al.
Chemical science, 10(9), 2623-2632 (2019-04-19)
Single-site Ir(CO)2 complexes bonded to high-surface-area metal oxide supports, SiO2, TiO2, Fe2O3, CeO2, MgO, and La2O3, were synthesized by chemisorption of Ir(CO)2(acac) (acac = acetylacetonate) followed by coating with each of the following ionic liquids (ILs): 1-n-butyl-3-methylimidazolium tetrafluoroborate, [BMIM][BF4], 1-n-butyl-3-methylimidazolium
Selective Hydrogenation of 1, 3-Butadiene to 1-Butene by Pd (0) Nanoparticles Embedded in Imidazolium Ionic Liquids
Umpierre AP, et al.
Advanced Synthesis & Catalysis, 347(10), 1404-1412 (2005)
Hydrogenation of 1, 3-butadiene and of crotonaldehyde over highly dispersed Au catalysts
Okumura M, et al.
Catalysis Today, 74(3-4), 265-269 (2002)
Ultimately Specific 1, 4-cis Polymerization of 1, 3-Butadiene with a Novel Gadolinium Catalyst
Kaita S, et al.
Macromolecular Rapid Communications, 24(2), 179-184 (2003)
A short enantioselective pathway for the synthesis of the anti-influenza neuramidase inhibitor oseltamivir from 1, 3-butadiene and acrylic acid
Yeung Y-Y, et al.
Journal of the American Chemical Society, 128(19), 6310-6311 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service