70400
2-Naphthoic acid
≥97.0% (GC)
Synonym(s):
2-Naphthalenecarboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C10H7CO2H
CAS Number:
Molecular Weight:
172.18
Beilstein:
972039
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥97.0% (GC)
form
powder
mp
180-185 °C
185-187 °C (lit.)
SMILES string
OC(=O)c1ccc2ccccc2c1
InChI
1S/C11H8O2/c12-11(13)10-6-5-8-3-1-2-4-9(8)7-10/h1-7H,(H,12,13)
InChI key
UOBYKYZJUGYBDK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
V Krishnakumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 70(1), 201-209 (2007-09-08)
The solid phase mid FTIR and FT Raman spectra of 2-naphthoic acid (NA) and 6-bromo-2-naphthoic acid (BNA) have been recorded in the regions 4000-400 cm(-1) and 3500-100 cm(-1), respectively. The fundamental vibrational frequencies and intensities of the vibrational bands were
M J Melancon et al.
Drug metabolism and disposition: the biological fate of chemicals, 10(2), 128-133 (1982-03-01)
Urine was collected from four female rats for 3 days after two subcutaneous injections with 0.3 mg of 2-methyl[8-14C]naphthalene per kg. Of the 14C injected, 55% was found in the urine. The urine was solvent-fractionated into a toluene fraction (4.9%
Hui-Kai Huang et al.
Journal of industrial microbiology & biotechnology, 39(1), 55-62 (2011-06-18)
A heat-labile phenolic acid decarboxylase from Candida guilliermondii (an anamorph of Pichia guilliermondii) was purified to homogeneity by simple successive column chromatography within 3 days. The molecular mass was 20 kDa by sodium dodecyl sulfate-polyacrylamide gel electrophoresis and 36 kDa
Blaise Mathias Costa et al.
Neuropharmacology, 62(4), 1730-1736 (2011-12-14)
Over-activation of N-methyl-d-aspartate (NMDA) receptors is critically involved in many neurological conditions, thus there has been considerable interest in developing NMDA receptor antagonists. We have recently identified a series of naphthoic and phenanthroic acid compounds that allosterically modulate NMDA receptors
L J Fitzgerald et al.
Acta crystallographica. Section C, Crystal structure communications, 49 ( Pt 11), 1952-1958 (1993-11-15)
The structures of 1-naphthoic acid and 2-naphthoic acid have been investigated in order to determine the degree of disorder of the carboxylic acid groups. 1-Naphthoic acid was found to be completely ordered with C--O bond lengths of 1.214 (3) and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service