Skip to Content
Merck
All Photos(2)

Key Documents

663077

Sigma-Aldrich

(5R)-(+)-2,2,3-Trimethyl-5-benzyl-4-imidazolidinone dichloroacetic acid

97%

Synonym(s):

(5R)-2,2,3-Trimethyl-5-phenylmethyl-4-imidazolidinone dichloroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20Cl2N2O3
CAS Number:
Molecular Weight:
347.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

122-126 °C

functional group

carboxylic acid
chloro
phenyl

SMILES string

OC(=O)C(Cl)Cl.CN1C(=O)[C@@H](Cc2ccccc2)NC1(C)C

InChI

1S/C13H18N2O.C2H2Cl2O2/c1-13(2)14-11(12(16)15(13)3)9-10-7-5-4-6-8-10;3-1(4)2(5)6/h4-8,11,14H,9H2,1-3H3;1H,(H,5,6)/t11-;/m1./s1

InChI key

YZRAPAXOLQHZSE-RFVHGSKJSA-N

Application

(5R)-(+)-2,2,3-Trimethyl-5-benzyl-4-imidazolidinone dichloroacetic acid is a Macmillan imidazolidinone organocatalyst that can be used in the enantioselective organocatalytic α-fluorination of aldehydes and cyclic ketones.

Legal Information

U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa D Beeson et al.
Journal of the American Chemical Society, 127(24), 8826-8828 (2005-06-16)
The first direct enantioselective catalytic alpha-fluorination of aldehydes has been accomplished. The use of enamine catalysis has provided a new organocatalytic strategy for the enantioselective fluorination of aldehydes to generate alpha-fluoro aldehydes, an important chiral synthon for medicinal agent synthesis.

Articles

In collaboration with Materia, Inc., we are pleased to offer six imidazolidinone OrganoCatalysts™.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service