Skip to Content
Merck
All Photos(1)

Key Documents

647950

Sigma-Aldrich

3-Quinolineboronic acid pinacol ester

95%

Synonym(s):

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18BNO2
CAS Number:
Molecular Weight:
255.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

form

solid

mp

56-60 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2cnc3ccccc3c2

InChI

1S/C15H18BNO2/c1-14(2)15(3,4)19-16(18-14)12-9-11-7-5-6-8-13(11)17-10-12/h5-10H,1-4H3

InChI key

ARRJAONCYUAPJR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott McAuley et al.
Cell chemical biology, 26(9), 1274-1282 (2019-07-08)
Common approaches to antibiotic discovery include small-molecule screens for growth inhibition in target pathogens and screens for inhibitors of purified enzymes. These approaches have a shared intent of seeking to directly target a vital Achilles heel in a pathogen of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service