637319
Samarium(III) oxide
nanopowder, <100 nm particle size (BET), ≥99% trace metals basis
Synonym(s):
Samaria
About This Item
Recommended Products
Assay
≥99% trace metals basis
form
nanopowder
reaction suitability
reagent type: catalyst
core: samarium
surface area
<10 m2/g
particle size
<100 nm (BET)
density
8.35 g/mL at 25 °C (lit.)
SMILES string
O=[Sm]O[Sm]=O
InChI
1S/3O.2Sm
InChI key
PRCWVHVINXAFRG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Magnetic materials permeate numerous daily activities in our lives. They are essential components of a diversity of products including hard drives that reliably store information on our computers, decorative magnets that keep the shopping list attached to the refrigerator door, electric bicycles that speed our commute to work, as well as wind turbines for conversion of wind energy to electrical power.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service