Skip to Content
Merck
All Photos(1)

Key Documents

630209

Sigma-Aldrich

Poly(fluorescein isothiocyanate allylamine hydrochloride)

Poly(allylamine hydrochloride) : Fluorescein isothiocyanate 50:1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

solubility

H2O: soluble

λmax

494 nm

Looking for similar products? Visit Product Comparison Guide

General description

Poly(allylamine hydrochloride) : Fluorescein isothiocyanate 50:1

Other Notes

Polymer was made using Poly(allylamine hydrochloride), Mw ca. 56,000

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bugra Ayan et al.
Lab on a chip, 16(22), 4366-4372 (2016-11-02)
On-chip microparticle and cell coating technologies enable a myriad of applications in chemistry, engineering, and medicine. Current microfluidic coating technologies often rely on magnetic labeling and concurrent deflection of particles across laminar streams of chemicals. Herein, we introduce an acoustofluidic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service