All Photos(1)
About This Item
Linear Formula:
NCC6H4ZnI
CAS Number:
Molecular Weight:
294.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
concentration
0.5 M in THF
density
0.998 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
I[Zn]c1cccc(c1)C#N
InChI
1S/C7H4N.HI.Zn/c8-6-7-4-2-1-3-5-7;;/h1-2,4-5H;1H;/q;;+1/p-1
InChI key
MWVOJRQBIBGPBW-UHFFFAOYSA-M
Application
3-Cyanophenylzinc iodide can be used:
- As a reagent/substrate in the palladium-catalyzed synthesis of 5-substituted 2-furaldehydes of biological importance.
- As a substrate in the di-functionalization of indenes via α-carbonylalkylarylation to yield α-carbonyl- alkylarylated indenes.
Legal Information
Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
1.0 °F - closed cup
Flash Point(C)
-17.22 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A convenient synthesis of 5-aryl-and 5-heteroaryl-2-furaldehydes by the cross-coupling reaction of organozincs
Kim S and Rieke RD
Tetrahedron Letters, 51(19), 2657-2659 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service