Skip to Content
Merck
All Photos(1)

Key Documents

416037

Sigma-Aldrich

Poly(acrylic acid, sodium salt) solution

average Mw ~15,000, 35 wt. % in H2O

Synonym(s):

PAA, Sodium polyacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CO2Na)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mw ~15,000

Quality Level

concentration

35 wt. % in H2O

refractive index

n20/D 1.405

density

1.25 g/mL at 25 °C

SMILES string

[Na]OC(=O)C=C

InChI

1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1

InChI key

NNMHYFLPFNGQFZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priscila Stona et al.
The journal of adhesive dentistry, 15(3), 221-227 (2013-04-06)
To examine the influence of 11.5% polyacrylic acid pretreatment on the interface and bond strength of self-adhesive resin cements (Maxcem Elite, RelyX Unicem, SeT) to dentin. Fifty-six third molars were randomly divided into seven groups: RelyX ARC as control (ARC)
Chunjiao Zhou et al.
Journal of nanoscience and nanotechnology, 13(7), 4627-4633 (2013-08-02)
Poly(acrylic acid) (PAA) coated-Fe3O4 superparamagnetic nano-composites were synthesized through a solvothermal technique by using cheap and environmental friendly iron salts and PAA. Each nano-composite was composed of many small primary nanocrystals. The as-synthesized products were characterized by X-ray diffraction (XRD)
Serhat Elcicek et al.
Experimental parasitology, 133(3), 237-242 (2012-12-15)
Polyacrylic acid (PAA) is one of the anionic synthetic polyelectrolytes and is used in various immunological and pharmaceutical applications. PAA has been used as adjuvant in veterinary vaccines, in particular. However, to our knowledge, there are no studies that document
Michael Dietzsch et al.
Langmuir : the ACS journal of surfaces and colloids, 29(9), 3080-3088 (2013-02-08)
Scale formation, the deposition of certain minerals such as CaCO3, MgCO3, and CaSO4·2H2O in industrial facilities and household devices, leads to reduced efficiency or severe damage. Therefore, incrustation is a major problem in everyday life. In recent years, double hydrophilic
Marguerite J Kutyła et al.
AAPS PharmSciTech, 14(1), 301-311 (2013-01-12)
The controlled release of diflunisal and fluconazole from tablets made of novel polymers, poly(acrylic acid) (PAA) crosslinked with either β-cyclodextrin (βCD) or hydroxypropyl-βCD (HPβCD), was investigated and Carbopol 934P (Carbopol) was used as a highly crosslinked PAA for comparison. Diflunisal

Articles

Recently, layer-by-layer (LbL) assembly has emerged as a versatile, gentle and, simple method for immobilization of functional molecules in an easily controllable thin film morphology.1,2 In this short review, we introduce recent advances in functional systems fabricated by using the mild, yet adaptable LbL technique.

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service