Proteins, mixture
Materiales
Analytical column
column
TSKgel Ether-5PW, 7.5 cm × 7.5 mm I.D., 10 μm particles (808641)
mobile phase
0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 1.8 M to 0 M in 60 min
flow rate
1 mL/min
detector
UV, 280 nm
Descripción
Legal Information
TSKgel is a registered trademark of Tosoh Corporation