MilliporeSigma
HPLC Analysis of Glutathione S-Transferase on TSKgel® G3000SWXL
HPLC Analysis of Glutathione S-Transferase on TSKgel® G3000SWXL application for HPLC
Materiales
Application
Referencia del producto
Descripción
Precios
Columna para HPLC TSKgel® G3000SWXL
phase diol, L × I.D. 30 cm × 7.8 mm, 5 μm particle size
CONDITIONS
column
TSKgel G3000SWXL, 30 cm × 7.8 mm I.D., 5 μm particles (808541)
mobile phase
0.3M sodium chloride/0.05M sodium dihydrogen phosphate, pH 7.0
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220nm and enzyme assay
injection
100μL crude glutathione S-transferase preparation from guinea pig liver (0.7mg)
Descripción
Legal Information
TSKgel is a registered trademark of Tosoh Corporation