重要文件
33183
Supelco
手动启盖器
for use with 20mm crimp seals, pkg of 1 ea
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41122001
描述
Hand decapper
包裝
pkg of 1 ea
相容性
for use with 20mm crimp seals
尋找類似的產品? 前往 產品比較指南
一般說明
Hand decapper is used for quick and easy removal of a 20mm aluminum seal.
從最近期的版本中選擇一個:
分析證明 (COA)
Lot/Batch Number
如果您需要協助,請聯絡 客戶支援
已經擁有該產品?
您可以在文件庫中找到最近購買的產品相關文件。
存取文件庫
文章
Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.
Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.