重要文件
27533
Supelco
预组装广口螺纹口样品瓶,带聚丙烯瓶盖(10-425 螺纹)
volume 2 mL, clear glass vial, PTFE/silicone/PTFE septum, pkg of 100 ea
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41122001
材料
PTFE/silicone/PTFE septum
black polypropylene cap
clear glass vial
特點
closure type black polypropylene hole cap
closure type screw top vial
包裝
pkg of 100 ea
外徑 × 高度 × 內徑
12 mm × 32 mm × 6 mm
容積
2 mL
接頭
thread for 10-425
尋找類似的產品? 前往 產品比較指南
一般說明
Vials are suitable basically to inject samples from an autosampler. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of the sample from the vials. Screw top clear glass vials are used on many autosamplers with a large opening of 6 mm. The threads on the larger opening vials are 10-425. Includes a black polypropylene hole cap and a PTFE/silicone/PTFE septum, suitable for use with most organic solvents (until pierced) and not recommended for use with chlorosilanes.
適合性
Autosampler compatibility:
PerkinElmer Autosystem
PE ISS-100, 200, Integral 4000 and Series 200
Shimadzu GC and LC Autosamplers
Spark Marathon, Midas, Triathlon
Thermo HPLC Spectra System
Waters Alliance
其他說明
内插管的直径因制造商而不同。只有下列产品号的内插管与此类广口样品瓶兼容。
  • 24719,24720 6×29mm 锥形底部玻璃内插管
  • 24721 6×29mm 带底部支架的玻璃内插管
  • 24722 6×29mm 带底部支架的聚丙烯内插管
  • 24717,24718 6×31mm 锥形底部玻璃内插管
  • 24715,24716 6×31mm 水平底部玻璃内插管
從最近期的版本中選擇一個:
分析證明 (COA)