重要文件
Z741756
Manifold
4-Place
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41100000
NACRES:
NB.22
材料
polysulfone
特點
autoclavable
Includes necessary tubing – 1/8” i.d. silicone
製造商/商標名
IBI ACCFL0007
尋找類似的產品? 前往 產品比較指南
一般說明
The manifold allows for the connection and simultaneous delivery of media to flow cells and for consolidation of effluent from cells to the waste bottle. Comes with 1/8” i.d. silicone tubing attached and is autoclavable and re-usable.
從最近期的版本中選擇一個:
分析證明 (COA)