重要文件
Z719595
Sorenson low binding standard tips
maximum volume 200 μL, pkg of 96 tips CellRack, sterile
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121600
NACRES:
NB.01
材料
colorless
natural tip
polypropylene
無菌
sterile
特點
barrier: no
包裝
case of 4800 ea (50 racks of 96 tips)
pkg of 96 tips CellRack
pack of 960 ea (10 racks of 96 tips)
製造商/商標名
Sorenson 15280T
最大體積
200 μL
容積
200 μL
尋找類似的產品? 前往 產品比較指南
相關類別
一般說明
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
法律資訊
Sorenson is a trademark of Sorenson BioScience, Inc.
從最近期的版本中選擇一個:
分析證明 (COA)