重要文件
EP5391070026
Eppendorf(R) Plate Adapter HeatSealer S100
for 96 and 384 well plates
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121815
NACRES:
NB.15
特點
for 96 and 384 well plates
製造商/商標名
Eppendorf® 5391070026 (Global)
尋找類似的產品? 前往 產品比較指南
特點和優勢
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
從最近期的版本中選擇一個:
分析證明 (COA)