重要文件
EP022822080
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-72-8, non aerosol tight, to hold, 72 x 0.5 mL reaction vessels
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41103909
特點
to hold 72 x 0.5 mL reaction vessels
fixed angle rotor
non aerosol tight
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 022822080 (NA)
技術
centrifugation: suitable
相容性
for use with centrifuge models Vacufuge Plus Concentrator
尋找類似的產品? 前往 產品比較指南
相關類別
一般說明
Selection of rotors for the Eppendorf Vacufuge Plus.
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG