重要文件
BR31578
BRAND® BLAUBRAND®量筒,有序号
capacity 2000 mL
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121800
包裝
pkg of 12 ea
製造商/商標名
BRAND 27452
高度
115 mm
容量
2000 mL
尋找類似的產品? 前往 產品比較指南
一般說明
Measuring cylinders tall form. Class A DE-M marking to DIN EN ISO 4788. Made from DURAN(R), calibrated to contain (TC, In). With spout and hexagonal glass base. Ring marks at major graduations. High contrast blue enamel graduations and inscription
法律資訊
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
從最近期的版本中選擇一個:
分析證明 (COA)