P83067
1 of 10
Sigma-Aldrich
317240
盐酸二甲双胍
49163
葡萄糖 溶液
HUMANPBMC-0002644
Frozen Human Normal PBMC
Millipore
SCR005
蛋白酶细胞分离液
BR705893
BRAND® Transferpette® S Pipette Starter Kit
WHAWB100020
Qiagen Cutting Mat
Supelco
PHR1084
二甲双胍 盐酸盐
WHAWB100040
Unicore Punch Kit
M0605000
BP1296
Metformin for system suitability
AccuSMART
for use with AccuSMART Reader
Bioprocessing Systems
Tangential Flow Filtration Holders & Systems
Ultrafiltration & Diafiltration
輸入產品批次/批號來搜索 分析證明 (COA)。在產品’s標籤上找到批次和批號,寫有 ‘Lot’或‘Batch’.。
產品編號
批次/批編號