Merck
MFGL02SF3
Durapore® 0.22 µm, Millipak® Final Fill灌装囊式过滤器
Millipak® Final Fill 20
登入查看組織和合約定價
About This Item
分類程式碼代碼:
23151806
材料
PVDF membrane
polysulfone housing
silicone O-ring
無菌
sterile; irradiated
產品線
Millipak® Final Fill 20
特點
hydrophilic
技術
sterile filtration: suitable
基質
Durapore®
孔徑
0.22 μm pore size
接頭
fitting (19 mm (3/4 in.) Sanitary Flange Inlet and Outlet)
尋找類似的產品? 前往 產品比較指南
法律資訊
Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany