1.67525
1 of 10
Supelco
1.67520
Dip tube (PE) for acids and bases in 200L PE-drums
901748
Dip tube
1.01114
Withdrawal system
1.67521
Dip tube (PVDF) for sulfuric acid in 200 L PE-drums
1.67526
Dip tube (PE) for acids and bases in 25L fassett
901919
Hand pump ball
Sigma-Aldrich
Z280348
PVC laboratory tubing
902335
1.67527
Dip tube (PVDF) for nitric acid and sulfuric acid
BR143255
BRAND®专用实验室管件
輸入產品批次/批號來搜索 分析證明 (COA)。在產品’s標籤上找到批次和批號,寫有 ‘Lot’或‘Batch’.。
產品編號
批次/批編號