跳轉至內容
Merck
全部照片(1)

重要文件

166901

Millipore

Fluid A

bottle of, ready-to-use, bottle volume 300 mL , filling volume

登入查看組織和合約定價


About This Item

agency

EP 2.6.1
JP 4.06
USP 71

品質等級

無菌

sterile; autoclaved

形狀

liquid

特點

closure type screw cap with septum
ready-to-use

包裝

bottle of

技術

sterile filtration: suitable

瓶容量

500 mL
300 mL , filling volume

應用

cosmetics
food and beverages
pharmaceutical
sterility testing

儲存溫度

2-25°C

適合性

nonselective for

一般說明

Fluid A is ageneral rinse buffer designed in compliance with the European Pharmacopeia (EP)2.6.1, United States Pharmacopeia (USP) 71 and Japanese Pharmacopeia (JP) 4.06.The product is a clear colorless liquid.
Each bottle has a screwcap with septum closure to reduce the risk of cross-contamination and optimizethe disinfection procedures.

應用

Fluid A isused to rinse aqueous solutions during sterility testing by membranefiltration. It is also used to dilute soluble solids for the same application andas a rinsing fluid for membrane filtration of non-sterile products.
The productis robust and suitable for:
  • Dissolvingor diluting samples,
  • Reconstitutingcommercial microorganism sample.
  • As a transport medium formicroorganisms.

儲存類別代碼

10 - Combustible liquids

水污染物質分類(WGK)

WGK 1

閃點(°F)

Not applicable

閃點(°C)

Not applicable


分析證明 (COA)

輸入產品批次/批號來搜索 分析證明 (COA)。在產品’s標籤上找到批次和批號,寫有 ‘Lot’或‘Batch’.。

已經擁有該產品?

您可以在文件庫中找到最近購買的產品相關文件。

存取文件庫

我們的科學家團隊在所有研究領域都有豐富的經驗,包括生命科學、材料科學、化學合成、色譜、分析等.

聯絡技術服務