推薦產品
AC/DC 輸入
5 - 400 mA (Max Wattage: 80W; Voltage Output: 10-300V (5V increments))
特點
output terminals: 4 pairs
touchscreen control; Dual orientation, auto rotating display
包裝
pkg of 1
製造商/商標名
Benchmark Scientific E2301-E-UK
相容性
with most brands of gel boxes
尋找類似的產品? 前往 產品比較指南
一般說明
The Accuris myVolt Touch is a dependable electrophoresis power supply that includes all of the performance and specifications needed for efficient seperations of nucleic acids and proteins. Perfect for agarose gels and protein seperations, the myVolt Touch incorporates some very unique user-friendly features: Touch-screen Control Panel: Setting up an electrophoresis run with the myVolt Touch couldn′t be easier... simply touch the parameter that you want to change and then touch "start" to begin the run.
Two sets of output jacks are available to connect up to 2 gel rigs and the instruments can securely be stacked for use with additional gel rigs.
Two sets of output jacks are available to connect up to 2 gel rigs and the instruments can securely be stacked for use with additional gel rigs.
法律資訊
myVolt is a trademark of Benchmark Scientific, Inc.
Global Trade Item Number
庫存單位 | GTIN |
---|---|
Z742606-1EA | 4061826162156 |