Merck
全部照片(3)
重要文件
Z530891
Sigma-Aldrich
Flexible PTFE adapter
top female joint: ST/NS 24/40, bottom male joint: ST/NS 24/40
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121815
NACRES:
NB.35
接合處
female joint: ST/NS top24/40
male joint: ST/NS bottom24/40
尋找類似的產品? 前往 產品比較指南
相關類別
一般說明
Used for the transfer of powders and liquids. Can be compressed to approximately 190 mm or expanded to 250 mm. Glass joints with flexible corrugated PTFE tubing is pliable enough to create an offset adapter.
其他說明
Picture is an example of male and female joint style.
從最近期的版本中選擇一個:
分析證明 (COA)