參數
>5 psi pressure
尋找類似的產品? 前往 產品比較指南
一般說明
The Aldrich SAFE-PURGE™ valves provide for safe, high-performance operation. Both the manual and automatic spring-loaded styles feature a durable PTFE metering valve.
- Quick and efficient purging of reaction vessel which conserves expensive purge gases
- Safely vents toxic gases from built-in oil bubbler
Used with purge gas pressure greater than 5 psi. Should any oil pass the check valve, the spring-loaded plug will prevent any oil or air from entering the system.
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SAFE-PURGE is a trademark of Sigma-Aldrich Co. LLC
Global Trade Item Number
庫存單位 | GTIN |
---|---|
Z225339-1EA | 4061826227923 |