重要文件
TEM-G400F1-AU
Sigma-Aldrich
Gilder Finder Grids G400F1
Gold, size 400 mesh, vial of 100
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41111761
NACRES:
MA.24
尺寸:
400 mesh
包裝
vial of 100
尺寸
400 mesh
尋找類似的產品? 前往 產品比較指南
一般說明
Gilder Finder Grids G400F1. Gold grids, 400 mesh. The block of 1152 (32x36) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each small block of 36 (6x6) cells has a central number from 1 to 32 for identification purposes.
從最近期的版本中選擇一個:
分析證明 (COA)