重要文件
BR4630230
BRAND® Dispensette® S 有机定量瓶口分液器
volume 5 mL, without recirculation valve
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
特點
DE-M marking
without recirculation valve
製造商/商標名
BRAND 4630230
容積
5 mL
接頭
(GL 28/S 28)
GL 32
GL 38
S 40
尋找類似的產品? 前往 產品比較指南
一般說明
Dispensette S Organic is ideal for dispensing of organic solvents and acids
特點和優勢
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
其他說明
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
從最近期的版本中選擇一個:
分析證明 (COA)
Lot/Batch Number
抱歉,我們目前沒有在線上提供本產品的COA
如果您需要協助,請聯絡 客戶支援
已經擁有該產品?
您可以在文件庫中找到最近購買的產品相關文件。
存取文件庫
條款
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.