重要文件
BR4600220
BRAND®Dispensette® S 固定容量瓶口分液器
volume 2 mL, without recirculation valve
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.43
特點
DE-M marking
without recirculation valve
製造商/商標名
BRAND 4600220
容積
2 mL
接頭
(GL 28/S28)
GL 32
GL 38
S 40
尋找類似的產品? 前往 產品比較指南
一般說明
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
特點和優勢
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
其他說明
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
從最近期的版本中選擇一個:
分析證明 (COA)