重要文件
BMSBSH50022B
Sigma-Aldrich
myBlock ll digital dry bath with 2 Quick-Flip blocks for tube
strips and plates, AC/DC input 115 V AC, US 3-pin plug
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41102404
NACRES:
NB.76
製造商/商標名
Benchmark Scientific BSH5002-2B
參數
+5 °C temp. range (+105 °C)
5-105 °C temp. range
容量
, holds 2 blocks
尋找類似的產品? 前往 產品比較指南
一般說明
myBlock series is a "next generation" product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature.
MyBlock′s transparent lid helps to promote better uniformity and minimize temperature stratification in the block by maintaining a warm air jacket layer. The hinged lid can be flipped open (to the rear), or easily removed (lifted off) to accommodate taller tubes.
  • Digital temperature control (set and walk away)
  • Hinged lid (improves uniformity & conserves energy)
相關產品
產品號碼
描述
訂價
從最近期的版本中選擇一個:
分析證明 (COA)