Skip to Content
Merck
All Photos(1)

Key Documents

59137

Supelco

SUPELCOSIL LC-DABS (3 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOSIL LC-DABS HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm

manufacturer/tradename

SUPELCOSIL

extent of labeling

12.3% carbon loading

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

silica gel, high purity, spherical particle platform

matrix active group

C18 (octadecyl) phase

particle size

3 μm

pore size

120 Å

application(s)

food and beverages

compatibility

for use with Dabsyl-AA

separation technique

reversed phase

General description

SUPELCOSIL LC-DABS columns feature a specially treated and tested octadecylsilane bonded phase, for reversed-phase separations of precolumn derivatized dabsyl amino acids. More than 30 amino acids can be separated in less than one hour.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sainath Babu et al.
Toxicology mechanisms and methods, 25(5), 410-416 (2015-05-30)
We have developed a simple, reversed-phase high-performance liquid chromatography (RP-HPLC) method for the determination of bisphenol A (BPA) in thermal paper cash register receipts (CRs). The method is suitable for analysis of other types of bisphenols and it involves an
K K Peh et al.
International journal of pharmaceutical compounding, 4(3), 229-231 (2000-05-01)
A simple and selective high-performance liquid chromatography (HPLC) method using ultraviolet detection was developed for simultaneous determination of fusidic acid and betamethasone dipropionate in a cream formulation. A Supelcosil LC18 column was used for chromatographic separation. The mobile phase consisted
Vijay V Upreti et al.
Biomedical chromatography : BMC, 17(6), 385-390 (2003-09-19)
A simple and sensitive high-performance liquid chromatography (HPLC) method has been developed and validated for the determination of DRF-1042, a novel orally active camptothecin (CPT) analog, in human plasma. The sample preparation was a simple deproteinization with acidified methanol yielding
E M Koves
Journal of chromatography. A, 692(1-2), 103-119 (1995-02-10)
A comprehensive approach to the analysis for many drugs in postmortem blood and biological fluids using high-performance liquid chromatography and diode array detection has been developed. To reduce the likelihood of co-eluting interference components of postmortem blood or other drugs
S Djabarouti et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 799(1), 165-172 (2003-12-09)
The aim of this study was to develop a specific and sensitive high-performance liquid chromatographic (HPLC) assay for the determination of levofloxacin in human plasma, bronchoalveolar lavage and bone tissues. The sample extraction was based on a fully automated liquid-solid

Protocols

Amino Acid Analysis is the suitable tool for precise determination of protein quantities but also provides detailed information regarding the relative amino.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service