Skip to Content
Merck
All Photos(1)

Key Documents

56266-U

Adjustable Column Endpiece (Flow Adapter)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
SB.52

feature

adjustable

manufacturer/tradename

Omnifit 6165

technique(s)

LPLC: suitable

Looking for similar products? Visit Product Comparison Guide

Application

Plunger allows 80 mm of height adjustment, with a fine control that minimizes the risk of disturbance to the packed bed.
8 cm maximum adjustment.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service