Skip to Content
Merck
All Photos(1)

Key Documents

4S8231

Supelco

EPA Phthalate Esters Mix

certified reference material, 2000 μg/mL each component in hexane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 606,806,8061,8270

description

Separate Source

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 × 1 mL

concentration

2000 μg/mL each component in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Recommended products

This product is being deleted and is being replaced by product 48231

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Bis(2-ethylhexyl) phthalate
    Benzyl butyl phthalate
    Dibutyl phthalate
    Di-n-octyl phthalate
    Diethyl phthalate
    Dimethyl phthalate

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srimurali Sampath et al.
Environmental pollution (Barking, Essex : 1987), 221, 407-417 (2016-12-17)
Usage of phthalates as plasticizers has resulted in worldwide occurrence and is becoming a serious concern to human health and environment. However, studies on phthalates in Indian atmosphere are lacking. Therefore, we studied the spatio-temporal trends of six major phthalates
Shivani et al.
Chemosphere, 237, 124500-124500 (2019-09-25)
Deteriorating air quality with high levels of fine particulate matter (PM2.5) over National Capital Region (NCR) of India is one of the serious environmental and scientific issues. In this paper, PM2.5 samples were collected for 24 h twice or thrice a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service