Skip to Content
Merck
All Photos(1)

Key Documents

47369

Supelco

trans-1,3-Dichloropropene solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4Cl2
CAS Number:
Molecular Weight:
110.97
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C3H4Cl2/c4-2-1-3-5/h1-2H,3H2/b2-1+

InChI key

UOORRWUZONOOLO-OWOJBTEDSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo I Nikel et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 368(1616), 20120377-20120377 (2013-03-13)
Chlorinated pollutants are hardly biodegradable under oxic conditions, but they can often be metabolized by anaerobic bacteria through organohalide respiration reactions. In an attempt to identify bottlenecks limiting aerobic catabolism of 1,3-dichloroprop-1-ene (1,3-DCP; a widely used organohalide) in Pseudomonas pavonaceae
S R Yates et al.
Journal of environmental quality, 40(5), 1470-1479 (2011-08-27)
Soil fumigation is important for growing many fruits and vegetable crops, but fumigant emissions may contaminate the atmosphere. A large-scale field experiment was initiated to test the hypothesis that adding composted municipal green waste to the soil surface in an
Amit J Jhala et al.
Pest management science, 68(2), 225-230 (2011-07-29)
Preplant fumigation with methyl bromide (MeBr) has been used for control of soilborne pests in high-value annual, perennial and nursery crops, but is being phased out. In 2007 and 2008, research trials were conducted to evaluate the effects of surface
Pablo I Nikel et al.
Metabolic engineering, 15, 98-112 (2012-11-15)
Pseudomonas putida KT2440, a microbial cell factory of reference for industrial whole-cell biocatalysis, is unable to support biochemical reactions that occur under anoxic conditions, limiting its utility for a large number of relevant biotransformations. Unlike (facultative) anaerobes, P. putida resorts
J E Thomas et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 46(8), 655-661 (2011-08-03)
Soil fumigation to control pests and pathogens is an important part of current agricultural practice. A reduction in fumigant emissions is required to ensure worker safety and environment health. A field trial in Florida was conducted to investigate whether carbonating

Protocols

US EPA Method TO-17: GC Analysis of Volatiles on VOCOL® after Collection/Desorption using Air Toxics Tube

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service