24062
Untreated Fused Silica Tubing
L × I.D. 15 m × 0.32 mm
All Photos(1)
About This Item
Recommended Products
material
fused silica tubing
packaging
pkg of 1
technique(s)
gas chromatography (GC): suitable
L × I.D.
15 m × 0.32 mm
General description
Capillary columns have three distinctive layers namely, fused silica tube, protective coating and stationary phase. Fused silica tubing refers to tubing with a protective coating. Fused silica is the most commonly used material for capillary column tubing as it is a synthetic, quartz like glass containing less than 1ppm of metallic oxide impurities. Untreated fused silica is very active due to the presence of silanol groups.
Application
Untreated fused silica tubing is suitable for lining capillary column, used to support derivatized cyclodextrins as stationary phase immobilized on the column for enantiomeric separations via capillary electrophoresis, gas chromatography, or supercritical fluid chromatography.
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
Already Own This Product?
Derivatized cyclodextrins immobilized on fused-silica capillaries for enantiomeric separations via capillary electrophoresis, gas chromatography, or supercritical fluid chromatography.
Analytical Chemistry, 65 (8), 1114-1117 (1993)
Capillary GC Columns.
The Troubleshooting and Maintenance Guide for Gas Chromatographers (2007)