Skip to Content
Merck
All Photos(1)

Key Documents

22896

Chart Paper for Recorder

for use with Agilent/HP recorders, chart type, Z-fold JetPaper, W × L 8.5 in. × 11 in., package of 5 × 500 sheets

Synonym(s):

CH.PAPER Z-FOLD FOR HP3394/96 INTEGRATORS (PK.5)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
14111500

feature

chart type Z-fold JetPaper

packaging

package of 5 × 500 sheets

W × L

8.5 in. × 11 in.

compatibility

for use with Agilent/HP recorders
for use with PerkinElmer recorders
for use with Spectra Physics recorders
for use with Varian recorders
for use with Waters recorders

Looking for similar products? Visit Product Comparison Guide

General description

Main purpose of a recorder is to record electrical and non-electrical quantity as a function of time. Graphic recorders use elementary rudiments like chart paper for displaying and storing the physical quantity and pen for marking the variation in physical quantity. Chart paper is passed through the recorder at constant rate. The recorder itself becomes a device for measuring retention time. A certain number of inches or centimetres of chart paper corresponds to a certain number of minutes.

Application

Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.

Other Notes

Equivalent to Manufacturer′s Chart No. 5062-3561.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjanie Fernando et al.
Cells, 9(10) (2020-10-18)
Alpha synuclein has been linked to both sporadic and familial forms of Parkinson's disease (PD) and is the most abundant protein in Lewy bodies a hallmark of Parkinson's disease. The function of this protein and the molecular mechanisms underlying its
John Kenkel
Analytical Chemistry for Technicians, 366-366 (1994)
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service