22262
Valco® Precision Sampling Syringe, Series A, Removable needle
volume 250 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2.25 in. × 0.029 in. × 0.012 in.
Synonym(s):
SYRINGE,PS-A,250UL
All Photos(1)
About This Item
UNSPSC Code:
41122001
material
bevel tip stainless steel needle (point style 2)
description
Series A
feature
removable needle
packaging
pkg of 1 ea
manufacturer/tradename
VICI 010031
needle L × O.D. × I.D.
2.25 in. × 0.029 in. × 0.012 in.
volume
250 μL
General description
Series A - Removable Needle: This Pressure-Lok syringe allows storage of sample within the syringe. Rotary valve in the needle hub allows for sample pre-pressurization prior to injection.
- Integral sample storage valve - actuated by turning the nose piece
- Positive rear flange plunger stop - prevents plunger from blowing out of barrel at elevated pressure
- Modular construction - all components may be quickly disassembled for easy cleaning or replacement
- Leak-tight to 250 psi - liquids and gases
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)