Skip to Content
Merck
All Photos(2)

Key Documents

WH0003371M1

Sigma-Aldrich

Monoclonal Anti-TNC antibody produced in mouse

clone 3B4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HXB, Anti-TN, Anti-tenascin C (hexabrachion)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
clone:
3B4, monoclonal
application:
ELISA (i)
WB
species reactivity:
human
technique(s):
indirect ELISA: suitable
western blot: 1-5 μg/mL
citations:
3

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNC(3371)

Immunogen

TNC (NP_002151, 181 a.a. ~ 290 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KGPNCSEPECPGNCHLRGRCIDGQCICDDGFTGEDCSQLACPSDCNDQGKCVNGVCICFEGYAGADCSREICPVPCSEEHGTCVDGLCVCHDGFAGDDCNKPLCLNNCYN

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebekah L Hanson et al.
Journal of managed care pharmacy : JMCP, 19(1), 49-67 (2013-02-07)
Tumor necrosis factor (TNF)-alpha inhibitors and other biologic response modifiers (BRMs) are frequently used to treat a variety of inflammatory diseases. Use of these agents may increase risk of serious infections, malignancies, and other complications such as worsening symptoms of
Adeline Guais et al.
Gene, 374, 112-120 (2006-03-22)
In Drosophila, the RING finger protein d-Goliath was originally identified as a transcription factor involved in the embryo mesoderm formation [Bouchard, M.L., Cote, S., 1993. The Drosophila melanogaster developmental gene g1 encodes a variant zinc-finger-motif protein. Gene 125, 205-209]. In
W Böcker et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 23(11), 2437-2448 (2014-08-01)
In humans, glucocorticoid-induced osteoporosis is the most common cause of medication-induced osteoporosis. Recent clinical data suggest that glucocorticoid therapy increases the risk of vertebral fractures within a short treatment period. Therefore, this study aimed at investigating vertebral bone in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service