SRP6352
CD247 human
recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Synonym(s):
CD3-ZETA, CD3H, CD3Q, CD3Z, T-cell surface glycoprotein CD3 zeta chain, T3Z, TCRZ
About This Item
biological source
human
recombinant
expressed in E. coli
Assay
≥90% (SDS-PAGE)
form
liquid
mol wt
15.4 kDa (136 aa, 52-164 aa + His Tag)
packaging
pkg of 50 μg
concentration
0.5 mg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... CD247(919)
General description
Physical form
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service