Skip to Content
Merck
All Photos(1)

Key Documents

SRP6178

Sigma-Aldrich

TACE human

recombinant, expressed in insect cells, ≥90% (SDS-PAGE)

Synonym(s):

ADAM17, CD156b, Disintegrin and metalloproteinase domain-containing protein 17, Snake venom-like protease, TNF-alpha convertase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in insect cells

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

mol wt

62 kDa

packaging

pkg of 20 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ADAM17(6868)

General description

A disintegrin and metallopeptidase domain 17 (ADAM17) belongs to ADAM family of genes, which has a characteristic cysteine rich region called disintegrin and a metalloproteinase domain. This gene is located on chromosome 2, spans 55kb and has 19 exons. In adult humans, ADAM17 is expressed in multiple tissues such as, heart, muscle, kidney, placenta, pancreas, thymus, small intestine, ovaries, testes and prostate. In fetus, it is expressed in brain, lung, liver and kidney.

Biochem/physiol Actions

ADAMs (A disintegrin and metallopeptidases) are called ectodomain sheddases and ADAM17 is an angiotensin-converting enzyme 2 (ACE2) sheddase. It also processes and sheds multiple cell surface molecules such as, adhesion molecules and cytokines. It produces the soluble form of tumor necrosis factor α (TNF-α), along with the processing of I-CAM (cell adhesion molecule), V-CAM, IL-6R, L-selectin etc. These in turn play a role in development, regeneration and inflammation of the circulatory system. Polymorphism rs12692386 in ADAM17 gene is associated with an increased susceptibility to abdominal aortic aneurysm. This gene is also up-regulated in myocardial infarction patients, as well as in atherosclerosis patients. It also plays a role in the pathophysiology of ischemic strokes (IS). ADAM17 aids in tumor progression by facilitating tumor proliferation and invasion, as in the case of non-small cell lung cancer (NSCLC).

Physical form

The untagged rh-TACE (TNF-alpha converting enzyme) is supplied as a lyophilized powder.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

With sterile deionized water at 0.1 μg protein/μL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential surface expression of ADAM10 and ADAM17 on human T lymphocytes and tumor cells.
Ebsen H, et al.
PLoS ONE, 8(10) (2013)
You Li et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 33(5), 1426-1438 (2014-05-24)
Accumulating evidence suggests that the principal TNF-α converting enzyme, a disintegrin and metalloproteinase 17 (ADAM17), is involved in the development of human abdominal aortic aneurysm (AAA). However, the association between ADAM17 gene polymorphisms and AAA has not been explored. The
You Li et al.
Journal of atherosclerosis and thrombosis, 21(8), 878-893 (2014-04-15)
Stroke is a leading cause of death and disability worldwide. Most ischemic strokes (IS) are caused by atherosclerosis. Recently, the pivotal role of ADAM17 in atherosclerosis has been thoroughly addressed. However, the association between ADAM17 and IS has not yet
Xiaohong Lv et al.
Molecular medicine reports, 9(5), 1935-1940 (2014-03-15)
A disintegrin and metalloprotease (ADAM) 17 has been implicated in the tumor progression of various types of solid tumor; however, little is known about its role in non-small cell lung carcinoma (NSCLC). The present study evaluated whether the downregulation of
Qin Xu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(8), 7575-7586 (2014-05-06)
Metalloproteinase activities of a disintegrin and metalloproteinase 17 (ADAM17), amphiregulin (AREG), extracellular matrix metalloproteinase inducer (EMMPRIN), and matrix metalloproteinases (MMPs) are involved in tumor biology. In patients with uterine cervical carcinoma, the expression and prognostic significance of ADAM17 remain to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service