Skip to Content
Merck
All Photos(2)

Key Documents

SRP5118

Sigma-Aldrich

PDE4B (305-end), active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp686F2182, DPDE4, MGC126529, PDE4B5, PDEIVB

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

457-619 nmol/min·mg

mol wt

~78 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDE4B(5142)

General description

PDE4B is a member of the phosphodiesterase family of proteins that play a critical role in regulating intracellular levels of cAMP and has homology to the ′dunce′ gene of Drosophila. The ′dunce′ gene is one of several genes critical for normal learning and memory in the fly. Altered activity of PDE4B has been associated with schizophrenia and bipolar affective disorder. DISC1 interacts with the UCR2 domain of PDE4B and elevation of cellular cAMP leads to dissociation of PDE4B from DISC1 and in increase in PDE4B activity. PDE4B gene itself does not link to major depressive disorder (MDD) but elevated mRNA levels of PDE4B have been implicated in the pathophysiology of MDD.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shusuke Numata et al.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics, 150B(4), 527-534 (2008-09-12)
The phosphodiesterase 4B (PDE4B) interacts with disrupted-in-schizophrenia 1 (DISC1), which is a known genetic risk factor for schizophrenia, bipolar disorder and major depressive disorder (MDD). PDE4B is also important in the regulation of cAMP signaling, a second messenger implicated in
J Kirsty Millar et al.
Science (New York, N.Y.), 310(5751), 1187-1191 (2005-11-19)
The disrupted in schizophrenia 1 (DISC1) gene is a candidate susceptibility factor for schizophrenia, but its mechanistic role in the disorder is unknown. Here we report that the gene encoding phosphodiesterase 4B (PDE4B) is disrupted by a balanced translocation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service