SML0641
BAM7
≥98% (HPLC)
Synonym(s):
4-(2-(2-Ethoxyphenyl)hydrazono)-3-methyl-1-(4-phenylthiazol-2-yl)-1H-pyrazol-5(4H)-one, BAX activator molecule 7
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
≥98% (HPLC)
form
powder
color
light orange to dark orange
solubility
DMSO: 2 mg/mL, clear (warmed)
storage temp.
2-8°C
Biochem/physiol Actions
BAM7 is a selective activator of BAX, a proapoptotic member of the BCL-2 protein family. BAM7 binds directly to the BAX trigger site, a distinct BH3 binding site that regulates BAX activation, inducing BAX oligomerization, which enables the release of apoptogenic factors that result in cell death. BAM7 is selective for this previously unknown BH3-binding groove on the N-terminal face of BAX.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Developmental cell, 53(6), 627-645 (2020-06-07)
Mitochondrial outer membrane permeabilization (MOMP) is a core event in apoptosis signaling. However, the underlying mechanism of BAX and BAK pore formation remains incompletely understood. We demonstrate that mitochondria are globally and dynamically targeted by endolysosomes (ELs) during MOMP. In
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service