Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501371

Sigma-Aldrich

Anti-Histone H2A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

H2A.1b, H2A/c, H2A/d, H2A/i, H2A/n

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IHC
clone:
polyclonal
species reactivity:
mouse, rat, human
citations:
4
technique(s):
ELISA: 1:1000
immunohistochemistry: 1:50-1:100

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 13 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... H2AFZ(3015)

General description

Anti-Histone H2A antibody detects endogenous levels of total Histone H2A protein.
Histone H2A(histone family member Z) is located on human chromosome 4q23. It contains two isoforms-H2A.Z.1 and H2A.Z.2.

Immunogen

The antiserum was produced against synthesized peptide derived from human Histone H2A.

Immunogen Range: 1-50

Biochem/physiol Actions

Histone H2A(histone family member Z) participates in multiple epigenetic regulations and carcinogenesis. It also participates in embryonic stem (ES) cell biology. It regulates the expression of genes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Post-translational modifications of the histone variant H2AZ
Sevilla A and Binda O
Stem Cell Research, 12(1), 289-289 (2014)
Genetic complementation analysis showed distinct contributions of the N-terminal tail of H2A.Z to epigenetic regulations
Kusakabe M, et al.
Genes Cells, 21(2), 122-135 (2016)
Wioleta Justyna Omeljaniuk et al.
Scientific reports, 10(1), 13088-13088 (2020-08-06)
Pregnancy loss is a multidisciplinary problem which concerns researchers from the fields of medicine, epidemiology, psychology, and public health. The primary objective of the present study was to explain the potential role of neutrophil extracellular traps (NETs) in the process
A comprehensive view of the epigenetic landscape part I: DNA methylation, passive and active DNA demethylation pathways and histone variants.
Sadakierska-Chudy A
Neurotoxicity Research, 27(1), 84-97 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service