Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300695

Sigma-Aldrich

Anti-CDK4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CMM3, Anti-PSK-J3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~34 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(L-H-K-D-E)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDK4(1019)

General description

The antibody detects endogenous levels of total CDK4 protein.

Immunogen

Peptide sequence around aa. 295-299 (L-H-K-D-E), according to the protein NP_000066.1

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Cyclin-dependent kinase activity is regulated by T-loop phosphorylation (Thr172 in the case of CDK4), by the abundance of their cyclin partners, and by association with CDK inhibitors of the Cip/Kip or INK family of proteins . The inactive ternary complex of CDK4/cyclin D and p27 Kip1/Cip1 requires extracellular mitogenic stimuli for the release and degradation of p27, which affects progression through the restriction point and pRb-dependent entry into S-phase . The active complex of CDK4/cyclin D targets the retinoblastoma protein for phosphorylation, allowing the release of E2F transcription factors that activate G1/S-phase gene expression . In HeLa cells, upon UV irradiation, upregulation of p16 INK4A association with CDK4/cyclin D3 leads to a G2 delay, implicating CDK4/cyclin D3 activity in progression through the G2-phase of the cell cycle .

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohan Qu et al.
Journal of translational medicine, 13, 301-301 (2015-09-17)
PIWI proteins have important roles in tumorigenesis due to their interaction with piRNAs. Recent studies suggest that PIWI proteins affect prognosis of various cancers. In the present study, PIWI genes expression was assayed in non-small cell lung cancer (NSCLC). To
Chaitali Misra et al.
Human molecular genetics, 23(19), 5025-5035 (2014-05-27)
Mutations in GATA4 and TBX5 are associated with congenital heart defects in humans. Interaction between GATA4 and TBX5 is important for normal cardiac septation, but the underlying molecular mechanisms are not well understood. Here, we show that Gata4 and Tbx5
Guoyan Liu et al.
The Journal of pathology, 233(3), 308-318 (2014-03-08)
Ovarian carcinoma is the most lethal gynaecological malignancy. Better understanding of the molecular pathogenesis of this disease and effective targeted therapies are needed to improve patient outcomes. MicroRNAs play important roles in cancer progression and have the potential for use
Wen Pan et al.
Scientific reports, 4, 6812-6812 (2014-10-30)
Cytokines are soluble proteins that exert their functions by binding specific receptors. Many cytokines play essential roles in carcinogenesis and have been developed for the treatment of cancer. In this study, we identified a novel potential cytokine using immunogenomics designated
Jinhee Kim et al.
Biochimie, 105, 36-44 (2014-06-15)
Overexpression of the aromatase enzyme CYP19 has been implicated in the onset of estrogen-dependent breast carcinogenesis. Obacunone, a natural compound present in citrus fruits, has been demonstrated for various biological activities including anti-cancer and anti-inflammatory properties. In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service