Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300173

Sigma-Aldrich

Anti-phospho-PTK2B (pTyr402) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CADTK antibody produced in rabbit, Anti-CAKB antibody produced in rabbit, Anti-FADK2 antibody produced in rabbit, Anti-FAK2 antibody produced in rabbit, Anti-PTK2B protein tyrosine kinase 2 beta antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~116 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(D-I-YP-A-E)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr402)

Gene Information

human ... PTK2B(2185)

General description

Protein tyrosine kinase 2β (PTK2B) is a member of the focal adhesion kinase family which is expressed in the brain. The gene encoding it is localized on human chromosome 8.

Immunogen

Peptide sequence around phosphorylation site of tyrosine 402 (D-I-Y(p)-A-E), according to the protein PTK2B.

Biochem/physiol Actions

Protein tyrosine kinase 2 β (PTK2B) takes part in the β2-mediated signaling pathway and activates mitogen activated protein kinase pathway. The protein has been shown to be upregulated in many tumors. PTK2B also has a role in cell proliferation and migration. It regulates ion channels which are induced by calcium.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Involved in calcium induced regulation of ion channel and activation of the map kinase signaling pathway. May represent an important signaling intermediate between neuropeptide activated receptors or neurotransmitters that increase calcium flux and the downstream signals that regulate neuronal activity. Interacts with the SH2 domain of Grb2. May phosphorylate the voltage-gated potassium channel protein Kv1.2. Its activation is highly correlated with the stimulation of c-Jun N-terminal kinase activity. Involved in osmotic stress-dependent SNCA 'Tyr-125' phosphorylation.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiguo Shan et al.
Molecular medicine reports, 20(2), 1459-1467 (2019-06-08)
Colorectal carcinoma (CRC) is a common malignancy of the digestive tract. MicroRNA (miR)‑214 is considered a key hub that controls tumor networks; therefore, the effects of miR‑214 on CRC were examined and its target gene was investigated in this study.
Integrins Influence the Size and Dynamics of Signaling Microclusters in a Pyk2-dependent Manner.
Steblyanko M
The Journal of Biological Chemistry, 290, 11833-11842 (2015)
Thomas P Keeley et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 31(12), 5172-5183 (2017-08-02)
Intracellular O
Pyk2 promotes tumor progression in multiple myeloma.
Zhang Y
Blood, 124, 2675-2686 (2014)
Hiroshi Yabuki et al.
Cell transplantation, 27(6), 979-993 (2018-05-01)
Posttransplantation lung ischemia-reperfusion (IR) injuries affect both patient survival and graft function. In this study, we evaluated the protective effects of infused human multilineage-differentiating stress-enduring (Muse) cells, a novel, easily harvested type of nontumorigenic endogenous reparative stem cell, against acute

Global Trade Item Number

SKUGTIN
SAB4300173-100UG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service