Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500780

Sigma-Aldrich

Anti-CASKIN1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
WB
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
ELISA: suitable
western blot: suitable

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 157 kDa

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASKIN1(57524)

General description

CASK (Calcium/calmodulin-dependent serine protein kinase) interacting protein 1 (CASKIN1) is expressed in the neuronal cells. The N terminal region of Caskin1 have six ankyrin repeats, two sterile α motifs (SAM domains) and a single SH3 domain. The C-terminal region possesses a long proline-rich region. In human chromosome, the gene CASKIN1 is localized on 16p13.3.

Biochem/physiol Actions

CASK interacting protein 1 (CASKIN1) is a scaffold protein involved in synapses. Caskin1 interacts with Calcium/calmodulin-dependent serine protein kinase (Cask) in the CASK interaction domain (CID) adjacent to SAM domain. Caskin1 also interacts with Nck (non-catalytic region of tyrosine kinase adaptor protein 1)/Dock (dedicator of cytokinesis), neurexin2, synaptotagmin, and septin4. Caskin1 interacts with EphB1 (ephrinB ligands) through Nck adapter protein. The interaction phosphorylates the tyrosine residue in the SH3 domain and causes changes in the SH3 domain. Caskin1 forms a tripartite complex with Cask/caskin1/velis. Caskin1 competes with Munc-interacting protein 1 (Mint1) to form the tripartite complex. The tripartite complex binds to the cytoplasmic tail of neurexin, which is enriched in the presynaptic sites.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500780.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of neuronal complexes in human X-linked brain diseases
Laumonnier F, et al.
American Journal of Human Genetics, 80(2), 205-220 (2007)
The molecular basis of the Caskin1 and Mint1 interaction with CASK
Stafford RL, et al.
Journal of Molecular Biology, 412(1), 3-13 (2011)
Tandem SAM domain structure of human Caskin1: a presynaptic, self-assembling scaffold for CASK
Stafford RL, et al.
Structure, 19(12), 1826-1836 (2011)
Szabolcs Pesti et al.
Cell communication and signaling : CCS, 10(1), 36-36 (2012-11-28)
Scaffold proteins have an important role in the regulation of signal propagation. These proteins do not possess any enzymatic activity but can contribute to the formation of multiprotein complexes. Although scaffold proteins are present in all cell types, the nervous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service