Skip to Content
Merck
All Photos(1)

Key Documents

SAB2107781

Sigma-Aldrich

Anti-MADCAM1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MADCAM1(8174)

Immunogen

The immunogen for anti-MADCAM1 antibody: synthetic peptide derected towards the C terminal of human MADCAM1

Sequence

Synthetic peptide located within the following region: TTSPESPDTTSPESPDTTSQEPPDTTSPEPPDKTSPEPAPQQGSTHTPRS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Truffi et al.
International journal of nanomedicine, 15, 8537-8552 (2020-11-12)
Assessment of inflammatory bowel disease (IBD) currently relies on aspecific clinical signs of bowel inflammation. Specific imaging of the diseased bowel regions is still lacking. Here, we investigate mucosal addressin cell adhesion molecule 1 (MAdCAM-1) as a reliable and specific
Antoine P Fournier et al.
Science translational medicine, 12(560) (2020-09-11)
Mucosal tissues play critical roles in health and disease as the primary barrier between the external world and the inner body, lining the digestive, respiratory, urinary, mammary, and reproductive tracts. Clinical evaluation of mucosal tissues is currently performed using endoscopy
Rachel Y Gao et al.
Mucosal immunology, 14(2), 479-490 (2020-10-03)
Primary sclerosing cholangitis (PSC) is a progressive fibrosing cholestatic liver disease that is strongly associated with inflammatory bowel disease (IBD). PSC-associated IBD (PSC-IBD) displays a unique phenotype characterized by right-side predominant colon inflammation and increased risk of colorectal cancer compared
Tsuyoshi Sato et al.
Scientific reports, 10(1), 13221-13221 (2020-08-09)
Integrin activation is associated with conformational regulation. In this study, we developed a system to evaluate conformational changes in α4β7 integrin. We first inserted the PA tag into the plexin-semaphorin-integrin (PSI) domain of β7 chain, which reacted with an anti-PA
Anneleen Remmerie et al.
Immunity, 53(3), 641-657 (2020-09-06)
Metabolic-associated fatty liver disease (MAFLD) represents a spectrum of disease states ranging from simple steatosis to non-alcoholic steatohepatitis (NASH). Hepatic macrophages, specifically Kupffer cells (KCs), are suggested to play important roles in the pathogenesis of MAFLD through their activation, although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service