Skip to Content
Merck
All Photos(1)

Key Documents

SAB2106577

Sigma-Aldrich

Anti-TIMP2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

24 kDa

species reactivity

bovine, guinea pig, dog, horse, rat, sheep, human, rabbit, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIMP2(7077)
mouse ... Timp2(21858)

Immunogen

The immunogen for anti-TIMP2 antibody: synthetic peptide derected towards the C terminal of human TIMP2

Sequence

Synthetic peptide located within the following region: ECLWMDWVTEKSINGHQAKFFACIKRSDGSCAWYRGAAPPKQEFLDIEDP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Ozeki et al.
Oral diseases, 20(5), 505-513 (2013-08-02)
Matrix metalloproteinase (MMP)-3 expression increases after pulpectomy and accelerates angiogenesis in rat dental pulp by an uncharacterised mechanism. Odontoblasts, a major component of dental pulp, could represent a therapeutic target. We investigated whether MMP-3 activity is induced by cytokines and/or
Young-Rak Cho et al.
Oncology reports, 32(4), 1531-1536 (2014-08-12)
Broussonetia kazinoki (BK) has been used as a traditional medicine to improve vision, as well as for inflammatory and infectious diseases. In the present study, we investigated the effects and molecular mechanism of the ethanolic extract of BK on cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service