Skip to Content
Merck
All Photos(3)

Key Documents

SAB1408033

Sigma-Aldrich

Anti-KIAA1706 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

EEPD1, Endonuclease/exonuclease/phosphatase family domain containing 1, HSPC107, KIAA1706

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~62.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EEPD1(80820)

General description

KIAA1706 is also known as endonuclease-exonuclease-phosphatase family domain containing 1 (EEPD1). The gene is located on human chromosome 7p14.2. This gene contains two helix−hairpin−helix motifs at the N-terminal and an exonuclease/endonuclease/phosphatase domain at the C-terminal end. The protein is localized in the plasma membrane with a myristoylation site in its N terminus.

Immunogen

KIAA1706 (AAH65518.1, 1 a.a. ~ 569 a.a) full-length human protein.

Sequence
MGSTLGCHRSIPRDPSDLSHSRKFSAACNFSNILVNQERLNINTATEEELMTLPGVTRAVARSIVEYREYIGGFKKVEDLALVSGVGATKLEQVKFEICVSSKGSSAQHSPSSLRRDLLAEQQPHHLATAVPLTPRVNINTATPAQLMSVRGLSEKMALSIVDFRREHGPFRSVEDLVRMDGINAAFLDRIRHQVFAERSRPPSTHTNGGLTFTAKPHPSPTSLSLQSEDLDLPPGGPTQIISTRPSVEAFGGTRDGRPVLRLATWNLQGCSVEKANNPGVREVVCMTLLENSIKLLAVQELLDREALEKFCTELNQPTLPNIRKWKGPRGCWKAVVAEKPSNQLQKGAGYAGFLWDAAAGMELRDAGSQESSPSNGHGKLAGPSPYLGRFKVGSHDLTLVNLHLAALTLLGSENPSKNHSDGHRLASFAQTLQETLKGEKDVIILGDFGQGPDSNDYDILRKEKFHHLIPAHTFTNISTKNPQGSKSLDNIWISKSLKKVFTGHWAVVREGLTNPWIPDNWSWGGVASEHCPVLAEFYTEKDWSKKDAPRNGSGVALERSEANIKHER

Biochem/physiol Actions

Endonuclease-exonuclease-phosphatase family domain containing 1 (EEPD1), also known as KIAA1706, acts as a gatekeeper for the repair of stressed replication forks. It maintains the integrity of cell genome by developing safe homologous recombination fork repair pathway. Deficiency of this protein is associated with severe chromosomal abnormalities. The protein acts as a LXR-target gene, which maintains cholesterol efflux and activity of ATP-binding cassette transporter (ABCA1).

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EEPD1 rescues stressed replication forks and maintains genome stability by promoting end resection and homologous recombination repair
Wu Y, et al.
PLoS Genetics, 11(12), e1005675-e1005675 (2015)
Endonuclease EEPD1 is a gatekeeper for repair of stressed replication forks
Kim HS, et al.
The Journal of Biological Chemistry, jbc-M116 (2017)
EEPD1 is a novel LXR target gene in macrophages which regulates ABCA1 abundance and cholesterol efflux
Nelson JK, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 37(3), 423-432 (2017)
Synaptopodin and 4 novel genes identified in primary sensory neurons
Verpoorten N, et al.
Molecular and Cellular Neurosciences, 30(3), 316-325 (2005)
Array comparative genomic hybridization analysis of uterine leiomyosarcoma
Cho YL, et al.
Gynecologic Oncology, 99(3), 545-551 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service